Natriumthiosulfat solution_standard 0.1 mol/l (0.1 N) water
Lieferant: Thermo Fisher Scientific
Synonyme:
di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit
124270010
124270025
124270250
ACRO124270010EA
38.7
EUR
ACRO124270010
ACRO124270025
ACRO124270250
Natriumthiosulfat solution_standard 0.1 mol/l (0.1 N) water
Natriumthiosulfat
Formel:
Na₂S₂O₃ Molecular Weight: 158.11 g/mol Siedepunkt: 100 Schmelzpunkt: 48 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003499 CAS-Nummer: 7772-98-7 EINECS: 231-867-5 Merck Index: 12,08844 |
Spezifikation Testergebnisse
Appearance | Clear colorless Solution |
Titration Iodimetric | 0.0950 to 0.1050 N (20°C) |
Assay | ≥90 % (DAC) |
Loss on drying | ≤10.0 % |
Molecular weight | 600000 to 800000 Da |
Viscosity | 50 to 200 mPa.s |
Ash | ≤1.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...