N-(2-Hydroxyethyl)phthalimid 99%

Lieferant: Thermo Fisher Scientific

Synonyme: N-(2-Hydroxyethyl)phthalsäureimid, N-(2-Hydroxyethyl)-1,2-benzoldicarbonsäureimid

124920250
ACRO124920250EA 20.3 EUR
ACRO124920250
N-(2-Hydroxyethyl)phthalimid 99%
N-(2-Hydroxyethyl)phthalimid
Formel: C₁₀H₉NO₃
Molecular Weight: 191.19 g/mol
Schmelzpunkt: 126…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005903
CAS-Nummer: 3891-07-4
EINECS: 223-434-4

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 129 °C
HPLC ≥98.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR