(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 98%

Lieferant: Thermo Fisher Scientific

125130250
ACRO125130250EA 54.9 EUR
ACRO125130250
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 98%
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride

Appearance: White to yellowish-white Powder or crystals

Formel: C₇H₁₅NO₂·HCl
Schmelzpunkt: 145…155 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00012494
CAS-Nummer: 7517-19-3

Bestellen

Spezifikation Testergebnisse

Appearance White to yellowish-white Powder or crystals
Infrared spectrum Conforms
Specific optical rotation +18.5° to +21.5° (20°C, 589 nm) (c=4.5, methanol)
Titration with HClO4 ≥97.5 %
Residue after evaporation ≤0.005 %
UV 1 cm cell vs HPLC-grade water
Non-volatile matter ≤0.0003 %
Free acid (CH3COOH) ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR