L(+)-Lysin Monohydrochlorid 99%

Lieferant: Thermo Fisher Scientific

Synonyme: (S)-(+)-Lysin Monohydrochlorid, H-Lys-OH.HCl, (S)-(+)-2,6-Diaminohexansäure Hydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Hydrochlorid, L(+)-Lysin Hydrochlorid, (S)-(+)-2,6-Diaminohexansäure Monohydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Monohydrochlorid, (S)-(+)-Lysin Hydrochlorid

125220010 125221000 125222500 125220050
ACRO125220010EA 171 EUR
ACRO125220010 ACRO125221000 ACRO125222500 ACRO125220050
L(+)-Lysin Monohydrochlorid 99%
L(+)-Lysin Monohydrochlorid
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Schmelzpunkt: 263…264 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064564
CAS-Nummer: 657-27-2
EINECS: 211-519-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.4 % (105°C, 3 h)
Sulfated ash ≤0.1 %
Specific optical rotation +20.4° to +22.5° (20°C, 589 nm) (c=8, 6 N HCl)
Titration with HClO4 98.5 to 101.0 % (on dried substance)
Sulfate (SO4) ≤300 ppm
Iron (Fe) ≤0.005 %
Lead (Pb) ≤0.002 %
Nickel (Ni) ≤0.002 %
Insoluble matter ≤0.025 % (in water and ppt by NH4OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR