Methylphenylglyoxylat 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Methyl benzoylformate, Methyl 2-oxo-2-phenylacetate

126370250 126370050
ACRO126370250EA 79.5 EUR
ACRO126370250 ACRO126370050
Methylphenylglyoxylat 99%
Methylphenylglyoxylat
Formel: C₉H₈O₃
Molecular Weight: 164.1600 g/mol
Siedepunkt: 246…248 °C (760 mmHg)
Schmelzpunkt: 16 °C
Dichte: 1.15 g/cm³
Flammpunkt: 77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008443
CAS-Nummer: 15206-55-0
EINECS: 239-263-3

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5280 (20°C, 589 nm)
GC ≥98.5 %
Substance darkened by H2SO4 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR