2-Nitrozimtaldehyd 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
128610050
ACRO128610050EA
30.8
EUR
ACRO128610050
2-Nitrozimtaldehyd 98%
2-Nitrozimtaldehyd
Formel:
C₉H₇NO₃ Schmelzpunkt: 125…129 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00007188 CAS-Nummer: 1466-88-2 EINECS: 215-988-0 |
Spezifikation Testergebnisse
Appearance | Light yellow to yellow-brown Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 125 to 129 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...