(2-Nitrophenyl)-β-D-galactopyranosid (ONPG) 99%

Lieferant: Thermo Fisher Scientific

Synonyme: o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

128820010 128820050 128820250
ACRO128820010EA 46.2 EUR
ACRO128820010 ACRO128820050 ACRO128820250
(2-Nitrophenyl)-β-D-galactopyranosid (ONPG) 99%
(2-Nitrophenyl)-β-D-galactopyranosid (ONPG)
Formel: C₁₂H₁₅NO₈
Molecular Weight: 301.25 g/mol
Schmelzpunkt: 195 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00063255
CAS-Nummer: 369-07-3
EINECS: 206-716-1

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation -67° to -71° (20°C, 589 nm) (c=1, H2O)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR