L(+)-Ornithin Hydrochlorid 99%

Lieferant: Thermo Fisher Scientific

129560250 129561000 129565000
ACRO129560250EA 37.1 EUR
ACRO129560250 ACRO129561000 ACRO129565000
L(+)-Ornithin Hydrochlorid 99%
Formel: C₅H₁₂N₂O₂·HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00064562
CAS-Nummer: 3184-13-2
EINECS: 221-678-6

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Solubility (5 % in 1 N HCl) Clear colorless
Specific optical rotation +21° to +24° (20°C, 589 nm) (c=5, 1 N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR