1,10-Phenanthrolin Monohydrat 99+%, rein

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1,10-Phenanthroline hydrate

130130050 130130250 130131000 130135000
ACRO130130050EA 44.4 EUR
ACRO130130050 ACRO130130250 ACRO130131000 ACRO130135000
1,10-Phenanthrolin Monohydrat 99+%, rein
1,10-Phenanthrolin Monohydrat
Formel: C₁₂H₈N₂·1H₂O
Molecular Weight: 198.22 g/mol
Schmelzpunkt: 97…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149973
CAS-Nummer: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1 III
Merck Index: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Crystalline powder or needles
Infrared spectrum Conforms
Melting point 98 to 102 °C
Titration with HClO4 ≥99.0 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR