trans-2-Phenylcyclopropylamine hydrochloride 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
trans-2-Phenylcyclopropanamine hydrochloride, Tranylcypromine hydrochloride
130470010
130472500
130470050
ACRO130470010EA
111
EUR
ACRO130470010
ACRO130472500
ACRO130470050
trans-2-Phenylcyclopropylamine hydrochloride 97%
trans-2-Phenylcyclopropylamine hydrochloride
Appearance: White to light beige Powder and/or chunks
Formel:
C₉H₁₂ClN Schmelzpunkt: 162…169 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00063602 CAS-Nummer: 1986-47-6 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to light beige Powder and/or chunks |
Infrared spectrum | Conforms |
Titration Argentometric | ≥96.0 % |
HPLC | ≥96.0 % |
Residue after evaporation | ≤0.003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...