1,3-Phenylendiessigsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,2'-(1,3-Phenylene)diacetic acid
130500010
ACRO130500010EA
31.2
EUR
ACRO130500010
1,3-Phenylendiessigsäure 97%
1,3-Phenylendiessigsäure
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194,19 g/mol Schmelzpunkt: 173…177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004341 CAS-Nummer: 19806-17-8 EINECS: 243-332-3 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 171 to 179 °C |
Titration with NaOH | 96.0 to 104.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...