1-Phenyl-2-harnstoff 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
131020050
131021000
131022500
ACRO131020050EA
23.8
EUR
ACRO131020050
ACRO131021000
ACRO131022500
1-Phenyl-2-harnstoff 97%
1-Phenyl-2-harnstoff
Formel:
C₇H₈N₂O Schmelzpunkt: 143…149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007944 CAS-Nummer: 64-10-8 EINECS: 200-576-5 |
Spezifikation Testergebnisse
Appearance | Off-white to beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 143 to 149 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...