Propargylamin Hydrochlorid 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Propinylammoniumchlorid, Propargylammoniumchlorid, Monopropargylamin Hydrochlorid, Prop-2-ynylamin Hydrochlorid, 3-Aminopropin Hydrochlorid
131470010
131470100
ACRO131470010EA
39.6
EUR
ACRO131470010
ACRO131470100
Propargylamin Hydrochlorid 95%
Propargylamin Hydrochlorid
Formel:
C₃H₆ClN Schmelzpunkt: 175…182 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00012907 CAS-Nummer: 15430-52-1 EINECS: 239-440-5 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to beige to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 175 to 182 °C |
Titration Argentometric | 94 to 106 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...