3,5-Pyrazoledicarboxylic acid monohydrate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
131750050
131750250
ACRO131750050EA
53.3
EUR
ACRO131750050
ACRO131750250
3,5-Pyrazoledicarboxylic acid monohydrate 97%
3,5-Pyrazoledicarboxylic acid monohydrate
CAS No.: 3112-31-0
Appearance: White Crystalline powder
Formel:
C₅H₄N₂O₄·H₂O Schmelzpunkt: 292…295 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149323 CAS-Nummer: 303180-11-2 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Water | 8.0 to 11.0 % |
Titration with NaOH | 96.0 to 104.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...