2-Pyridinylessigsäurehydrochlorid 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(Pyridin-2-yl)acetic acid hydrochloride, 2-(Pyridin-2-yl)acetic acid HCl

131950050 131950250
ACRO131950050EA 54.5 EUR
ACRO131950050 ACRO131950250
2-Pyridinylessigsäurehydrochlorid 99%
2-Pyridylacetic acid hydrochloride

Appearance: White to beige Crystalline powder or crystals

Formel: C₇H₇NO₂
Schmelzpunkt: 135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012812
CAS-Nummer: 16179-97-8

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR