D(-)-Sorbit 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
D(-)-Glucit, (2R,3R,4R,5S)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Sorbitol, D(-)-Glucitol
132730010
132730025
132735000
132730050
132730100
132731000
ACRO132730010EA
64.8
EUR
ACRO132730010
ACRO132730025
ACRO132735000
ACRO132730050
ACRO132730100
ACRO132731000
D(-)-Sorbit 97%
D(-)-Sorbit
Formel:
HOH₂C(CH(OH))₄CH₂OH Molecular Weight: 182.17 g/mol Siedepunkt: 295 Schmelzpunkt: 95…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004708 CAS-Nummer: 50-70-4 EINECS: 200-061-5 Merck Index: 13,08797 |
|
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1.5 % |
Sulfated ash | ≤0.1 % |
HPLC | ≥96.0 % (on anhydrous substance) |
Specific optical rotation | +4.0° to +7.0° (20°C, 589 nm) (c=10, borate soln.) (on anhydrous substance) |
Lead (Pb) | ≤0.5 ppm |
Nickel (Ni) | ≤1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...