D(-)-Sorbit 97%

Lieferant: Thermo Fisher Scientific

Synonyme: D(-)-Glucit, (2R,3R,4R,5S)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Sorbitol, D(-)-Glucitol

132730010 132730025 132735000 132730050 132730100 132731000
ACRO132730010EA 64.8 EUR
ACRO132730010 ACRO132730025 ACRO132735000 ACRO132730050 ACRO132730100 ACRO132731000
D(-)-Sorbit 97%
D(-)-Sorbit
Formel: HOH₂C(CH(OH))₄CH₂OH
Molecular Weight: 182.17 g/mol
Siedepunkt: 295
Schmelzpunkt: 95…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004708
CAS-Nummer: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1.5 %
Sulfated ash ≤0.1 %
HPLC ≥96.0 % (on anhydrous substance)
Specific optical rotation +4.0° to +7.0° (20°C, 589 nm) (c=10, borate soln.) (on anhydrous substance)
Lead (Pb) ≤0.5 ppm
Nickel (Ni) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR