Aleuritic acid 95%

Lieferant: Thermo Fisher Scientific

133730250
ACRO133730250EA 157 EUR
ACRO133730250
Aleuritic acid 95%
Aleuritic acid

Appearance: White to beige Crystalline powder

Formel: C₁₆H₃₂O₅
Schmelzpunkt: 100…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135596
CAS-Nummer: 533-87-9

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥94 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR