1-Benzyl-3-methyl-4-piperidinon 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Benzyl-3-methyl-4-piperidone
134040250
ACRO134040250EA
156
EUR
ACRO134040250
1-Benzyl-3-methyl-4-piperidinon 99%
1-Benzyl-3-methylpiperidin-4-one
Appearance: Clear colorless to yellow Liquid
Formel:
C₁₃H₁₇NO Molecular Weight: 203,2800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044806 CAS-Nummer: 34737-89-8 UN: 2735 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5315 to 1.5335 (20°C, 589 nm) |
GC | ≥98 % |
Titration with HClO4 | 98.5 to 101.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...