9-Aminoacridin Hemihydrat 98%
Lieferant: Thermo Scientific
|
Achtung
|
134410010
ACRO134410010EA
62.5
EUR
ACRO134410010
9-Aminoacridin Hemihydrat 98%
9-Aminoacridin Hemihydrat
Formel:
C₁₃H₁₀N₂·0.5H₂O Schmelzpunkt: 230…235 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD28040496 CAS-Nummer: 65944-23-2 EINECS: 201-995-6 |
Spezifikation Testergebnisse
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | 96.0 to 104.0 % |
Specific optical rotation | +85° to +95° (20°C, 589 nm) (c=0.5, methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...