4-Acetyl-4-phenylpiperidine hydrochloride 99%

Lieferant: Thermo Scientific

134580050
ACRO134580050EA 156 EUR
ACRO134580050
4-Acetyl-4-phenylpiperidine hydrochloride 99%
4-Acetyl-4-phenylpiperidine hydrochloride

Appearance: Light yellow to beige Powder

Formel: C₁₃H₁₇NO
Schmelzpunkt: 232…234 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039037
CAS-Nummer: 10315-03-4

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR