Cyclohexylammoniumchlorid 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Cyclohexylamine Hydrochloride

134655000
ACRO134655000EA 90.3 EUR
ACRO134655000
Cyclohexylammoniumchlorid 99%
Cyclohexylamine hydrochloride

Appearance: White to beige Crystalline powder or crystals

Formel: C₆H₁₃N
Molecular Weight: 135,64 g/mol
Siedepunkt: 306 °C (760 mmHg)
Schmelzpunkt: 207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00050540
CAS-Nummer: 4998-76-9

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point ≥207 °C
Water ≤0.5 % (K.F.)
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR