2-Methyl-1-pyrroline 98%

Lieferant: Thermo Scientific
Gefahr

135070250
ACRO135070250EA 108 EUR
ACRO135070250
2-Methyl-1-pyrroline 98%
2-Methyl-1-pyrroline

Appearance: Clear colorless to yellow Liquid

Formel: C₅H₉N
Molecular Weight: 83,13 g/mol
Siedepunkt: 104…105 °C (760 mmHg)
Dichte: 0,878 g/cm³
Flammpunkt: 11 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00128806
CAS-Nummer: 872-32-2
UN: 1993
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR