1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one

135910010
ACRO135910010EA 101 EUR
ACRO135910010
1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone 97%
1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone

Appearance: Light yellow to beige Powder

Formel: C₁₂H₁₃N₃O
Molecular Weight: 215,26 g/mol
Schmelzpunkt: 195…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00044827
CAS-Nummer: 2147-83-3
UN: 3259
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 195 to 200 °C
Water ≤5 % (K.F.)
HPLC ≥96.0 %
UV Lambda max 280 to 284 nm
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR