4-[3-(Trifluoromethyl)phenyl]-4-piperidinol 97%

Lieferant: Thermo Fisher Scientific
Achtung

136980250
ACRO136980250EA 82.5 EUR
ACRO136980250
4-[3-(Trifluoromethyl)phenyl]-4-piperidinol 97%
4-[3-(Trifluoromethyl)phenyl]-4-piperidinol

Appearance: Light beige Crystalline powder

Formel: C₁₂H₁₄F₃NO
Schmelzpunkt: 91…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00044832
CAS-Nummer: 2249-28-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR