DL-Weinsäure 99.5%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (2R,3R)-rel-2,3-Dihydroxysuccinic acid, Racemic Acid

137875000 137870025 137871000
ACRO137875000EA 111 EUR
ACRO137875000 ACRO137870025 ACRO137871000
DL-Weinsäure 99.5%
DL-Tartaric acid

Appearance: White Crystalline powder

Formel: C₄H₆O₆
Molecular Weight: 150.09 g/mol
Schmelzpunkt: 200…206 °C
Flammpunkt: 210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151254
CAS-Nummer: 133-37-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 200 to 206 °C
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
Specific optical rotation -0.5° to +0.5° (20 °C, 589 nm) (c=20, H2O)
Titration with NaOH ≥99.4 %
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR