Tetramethylammoniumhydroxid 25% w/w methanol
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
138351000
138355000
138350025
ACRO138351000EA
39.3
EUR
ACRO138351000
ACRO138355000
ACRO138350025
Tetramethylammoniumhydroxid 25% w/w methanol
Tetramethylammonium hydroxide
Formel:
C₄H₁₃NO Molecular Weight: 91.15 g/mol Schmelzpunkt: –98 °C Dichte: 0.9 g/cm³ Flammpunkt: 6 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008280 CAS-Nummer: 75-59-2 EINECS: 200-882-9 UN: 3286 ADR: 3 II Merck Index: 14,09224 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Solution |
Titration with HCl | 24.5 to 26.0 % |
HPLC | ≥97.5 % (ELSD) |
Stabilizer | 2 % CaCO3 |
Specific optical rotation | +185° to +195° (20°C, 589 nm) (c=1, ethyl acetate) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...