Tri-O-acetyl-D-glucal 99%

Lieferant: Thermo Fisher Scientific

Synonyme: (-)-Tri-O-acetyl-D-glucal, 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tri-O-acetyl Ether, (2R,3S,4R)-2-(Acetoxymethyl)-3,4-dihydro-2H-pyran-3,4-diyl diacetate

139240250 139241000
ACRO139240250EA 78.3 EUR
ACRO139240250 ACRO139241000
Tri-O-acetyl-D-glucal 99%
Tri-O-acetyl-D-glucal

Appearance: White to off-white Crystalline powder

Formel: C₁₂H₁₆O₇
Molecular Weight: 272.25 g/mol
Schmelzpunkt: 51…53 °C
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00063253
CAS-Nummer: 2873-29-2

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (30°C, 15 h) (vacuum)
Sulfated ash ≤0.1 %
GC ≥98.5 %
Specific optical rotation -14.5° to -15.5° (20°C, 589 nm) (c=2, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR