L(-)-Tryptophan 99%

Lieferant: Thermo Fisher Scientific

140590050 140590250 140591000 140595000 140590010
ACRO140590050EA 27.3 EUR
ACRO140590050 ACRO140590250 ACRO140591000 ACRO140595000 ACRO140590010
L(-)-Tryptophan 99%
L(-)-Tryptophan
Formel: C₁₁H₁₂N₂O₂
Dichte: 1,34 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064340
CAS-Nummer: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929

Bestellen

Spezifikation Testergebnisse

Appearance White to yellowish-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Specific optical rotation -29.0° to -33.0° (20°C, 589 nm) (C=1, H2O)
Titration with HClO4 98.5 to 101.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR