3-(2-Hydroxyethyl)indol 97%

Lieferant: Thermo Fisher Scientific

Synonyme: β-Indol-3-ylethanol, Indol-3-ethanol, Tryptophol

140600050 140600010
ACRO140600050EA 71.5 EUR
ACRO140600050 ACRO140600010
3-(2-Hydroxyethyl)indol 97%
3-(2-Hydroxyethyl)indole
Formel: C₁₀H₁₁NO
Molecular Weight: 161.2 g/mol
Siedepunkt: 174 °C (2.7mbar)
Schmelzpunkt: 57…60 °C
Flammpunkt: 255 °C
MDL: MFCD00005659
CAS-Nummer: 526-55-6
EINECS: 208-393-2
Merck Index: 12,09930

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to brown Powder or crystals or flakes or crystalline mass
Infrared spectrum Conforms
Melting point 56 to 61 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR