(±)-3-Aminochinuclidin Dihydrochlorid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Aminoquinuclidine dihydrochloride, 3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

146150250
ACRO146150250EA 132 EUR
ACRO146150250
(±)-3-Aminochinuclidin Dihydrochlorid 98%
(±)-3-Aminochinuclidin Dihydrochlorid
Formel: C₇H₁₆Cl₂N₂
Molecular Weight: 199,1213 g/mol
Schmelzpunkt: 321…323 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00137395
CAS-Nummer: 6530-09-2
EINECS: 229-424-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR