1-(4-Nitrophenylazo)-2-naphthol, rein
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Para Red, 1-((4-Nitrophenyl)diazenyl)naphthalen-2-ol
146300250
ACRO146300250EA
113
EUR
ACRO146300250
1-(4-Nitrophenylazo)-2-naphthol, rein
1-(4-Nitrophenylazo)-2-naphthol
Formel:
C₁₆H₁₁N₃O₃ Molecular Weight: 293,28 g/mol Schmelzpunkt: 248…252 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003908 CAS-Nummer: 6410-10-2 EINECS: 229-093-8 |
Spezifikation Testergebnisse
Appearance | Red Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Elemental Analysis | 11.29 to 12.23 % (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...