Bis(4-hydroxyphenyl)sulfon 99.7%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4,4'-Dihydroxydiphenylsulfon, 4-Hydroxyphenylsulfon, 4,4'-Sulfonyldiphenol
146915000
ACRO146915000EA
74.2
EUR
ACRO146915000
Bis(4-hydroxyphenyl)sulfon 99.7%
Bis(4-hydroxyphenyl)sulfon
Formel:
C₁₂H₁₀O₄S Schmelzpunkt: >245 °C |
MDL:
MFCD00002350 CAS-Nummer: 80-09-1 EINECS: 201-250-5 |
Spezifikation Testergebnisse
Appearance | White to light pink Powder |
Infrared spectrum | Conforms |
Melting point | ≥245 °C |
Water | ≤0.5 % (K.F.) |
HPLC | ≥99.6 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...