Sudan I, rein

Lieferant: Thermo Fisher Scientific
Achtung

147060250 147061000
ACRO147060250EA 33.2 EUR
ACRO147060250 ACRO147061000
Sudan I, rein
Sudan I
Formel: C₁₆H₁₂N₂O
Siedepunkt: 444 °C (1013 hPa)
Schmelzpunkt: 131…133 °C
Dichte: 1,119 g/cm³ (151 °C)
MDL: MFCD00003911
CAS-Nummer: 842-07-9
EINECS: 212-668-2

Bestellen

Spezifikation Testergebnisse

Appearance Orange to red to brown Powder
Infrared spectrum Conforms
UV Lambda max at approximately 418 and 476 nm (in ethanol)
Specific extinction E (1%/1cm) ≥500 (at 476 nm, in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR