Cyclopentylmethanol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Cyclopentanmethanol

147150100
ACRO147150100EA 114 EUR
ACRO147150100
Cyclopentylmethanol 98%
Cyclopentylmethanol
Formel: C₆H₁₂O
Molecular Weight: 100,16 g/mol
Siedepunkt: 162 °C (1013 hPa)
Dichte: 0,92 g/cm³
Flammpunkt: 62 °C
MDL: MFCD00001384
CAS-Nummer: 3637-61-4
EINECS: 222-861-3

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4590 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR