3-Hydroxy-2-nitropyridin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Nitro-3-pyridinol

148100250 148101000
ACRO148100250EA 56.7 EUR
ACRO148100250 ACRO148101000
3-Hydroxy-2-nitropyridin 97%
3-Hydroxy-2-nitropyridine
Formel: C₅H₄N₂O₃
Molecular Weight: 140.1 g/mol
Schmelzpunkt: 67…72 °C
MDL: MFCD00006258
CAS-Nummer: 15128-82-2
EINECS: 239-191-2

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 72 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR