2-Acetylamino-4-methylthiazole-5-sulphonyl chloride 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Acetylamino-4-methyl-thiazole-5-sulphonyl chloride, 2-Acetamido-4-methyl-1,3-thiazole-5-sulphonyl chloride, 2-Acetamido-4-methylthiazole-5-sulphonyl chloride, 2-Acetamido-4-methyl-5-thiazolesulphonyl chloride
148360010
ACRO148360010EA
64.7
EUR
ACRO148360010
2-Acetylamino-4-methylthiazole-5-sulphonyl chloride 97%
2-Acetylamino-4-methylthiazole-5-sulphonyl chloride
Formel:
C₆H₇ClN₂O₃S₂ Molecular Weight: 254,72 g/mol Schmelzpunkt: 161…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005320 CAS-Nummer: 69812-29-9 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to brown Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...