Thiamin Hydrochlorid 98,5-101,5%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Thiaminiumchlorid, Antiberiberifaktor Hydrochlorid, antineuritisches Vitamin Hydrochlorid, Aneurin Hydrochlorid, Thiaminchlorid Hydrochlorid, Vitamin B1 Hydrochlorid

148990100 148991000 148990010
ACRO148990100EA 28.7 EUR
ACRO148990100 ACRO148991000 ACRO148990010
Thiamin Hydrochlorid 98,5-101,5%
Thiamin Hydrochlorid
Formel: C₁₂H₁₈Cl₂N₄OS
Schmelzpunkt: 250 °C (Zersetzung)
Dichte: 1,401 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012780
CAS-Nummer: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤5 % (K.F.)
Sulfated ash ≤0.1 %
Titration with HClO4 98.5 to 101.5 % (On anhydrous substance)
Sulfate (SO4) ≤300 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR