2-Methyl-2-propanthiol 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
tert-Butylmercaptan, tert-Butylthiol, 2-Mercapto-2-methylpropan
149020500
ACRO149020500EA
22.5
EUR
ACRO149020500
2-Methyl-2-propanthiol 99%
2-Methyl-2-propanthiol
Formel:
C₄H₁₀S Molecular Weight: 90.19 g/mol Siedepunkt: 64 Schmelzpunkt: –1.1 °C Dichte: 0.8 g/cm³ Flammpunkt: –24 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004857 CAS-Nummer: 75-66-1 EINECS: 200-890-2 UN: 2347 ADR: 3 II Merck Index: 13,01578 |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4220 to 1.4240 (20°C, 589 nm) |
GC | ≥98.5 % |
Dye content | ≥70 % |
UV Lambda max | 612 to 616 nm |
Suitability | (as track. dye for DNA seq) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...