Chromotrop 2B, rein

Lieferant: Thermo Fisher Scientific
Achtung

149750050 149750250
ACRO149750050EA 24.8 EUR
ACRO149750050 ACRO149750250
Chromotrop 2B, rein
Chromotrop 2B
Formel: C₁₆H₉N₃Na₂O₁₀S₂
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003951
CAS-Nummer: 548-80-1

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to greenish-brown to brown Powder
Infrared spectrum Conforms
Loss on drying ≤18 %
UV Lambda max 513 to 517 nm (c=0.1, H2O)
Specific extinction E (1%/1cm) ≥580 (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR