2,3-Butandionmonoxim 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Diacetylmonoxim
150371000
150375000
150370025
ACRO150371000EA
119
EUR
ACRO150371000
ACRO150375000
ACRO150370025
2,3-Butandionmonoxim 98%
2,3-Butandionmonoxim
Formel:
C₄H₇NO₂ Siedepunkt: 185…186 °C (760 mmHg) Schmelzpunkt: 74…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002116 CAS-Nummer: 57-71-6 EINECS: 200-348-5 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 74 to 77 °C |
Sulfated ash | ≤0.1 % |
Solubility | (5 % in water) Clear to slightly hazy |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...