2,3-Butandionmonoxim 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Diacetylmonoxim

150371000 150375000 150370025
ACRO150371000EA 119 EUR
ACRO150371000 ACRO150375000 ACRO150370025
2,3-Butandionmonoxim 98%
2,3-Butandionmonoxim
Formel: C₄H₇NO₂
Siedepunkt: 185…186 °C (760 mmHg)
Schmelzpunkt: 74…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002116
CAS-Nummer: 57-71-6
EINECS: 200-348-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 74 to 77 °C
Sulfated ash ≤0.1 %
Solubility (5 % in water) Clear to slightly hazy
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR