D(+)-Galactose 99+%

Lieferant: Thermo Fisher Scientific

Synonyme: (2R,3S,4S,5R)-Pentahydroxyhexanal

150610010 150611000 150615000 150610051 150610250
ACRO150610010EA 417 EUR
ACRO150610010 ACRO150611000 ACRO150615000 ACRO150610051 ACRO150610250
D(+)-Galactose 99+%
D(+)-Galactose
Formel: C₆H₁₂O₆
Molecular Weight: 180.16 g/mol
Siedepunkt: 527
Schmelzpunkt: 165…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151230
CAS-Nummer: 59-23-4
EINECS: 200-416-4
Merck Index: 13,04356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=3c02003b1d12c9bfd11c2e6faf5cc37d4325d208&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Water ≤0.3 % (K.F.)
Sulfated ash ≤0.1 % (on dry substance)
Solubility (10 % in water) Clear colorless
HPLC ≥99.0 % (ELSD)
Specific optical rotation +78° to +81.5° (20°C, 589 nm) (c=10, water, NH3) (after 30 minutes, on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR