Tetraethylammoniumchlorid Hydrat 99%

Lieferant: Thermo Fisher Scientific
Achtung

150900250 150901000 150905000
ACRO150900250EA 32 EUR
ACRO150900250 ACRO150901000 ACRO150905000
Tetraethylammoniumchlorid Hydrat 99%
Tetraethylammoniumchlorid Hydrat
Formel: C₈H₂₀ClN·xH₂O
MDL: MFCD00149992
CAS-Nummer: 56-34-8
EINECS: 200-267-5

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 % (on dry substance)
Loss on drying ≤10 %
Impurity ≤0.5 % Triethylamine hydrochloride


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR