Bromthymolblau, rein Indikator

Lieferant: Thermo Fisher Scientific

151360250 151361000 151365000
ACRO151360250EA 68.2 EUR
ACRO151360250 ACRO151361000 ACRO151365000
Bromthymolblau, rein Indikator
Bromthymolblau
Formel: C₂₇H₂₈Br₂O₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD00005872
CAS-Nummer: 76-59-5
EINECS: 200-971-2
Merck Index: 13,01430

Bestellen

Spezifikation Testergebnisse

Appearance Light pink to brown to purple Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Solubility (0.1 % in ethanol) Clear
Visual transition interval from pH 6.0 (yellow) to pH 7.6 (blue)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR