Bromthymolblau Natriumsalz, wasserlöslich, rein Indikator

Lieferant: Thermo Fisher Scientific

151370250 151371000
ACRO151370250EA 98.3 EUR
ACRO151370250 ACRO151371000
Bromthymolblau Natriumsalz, wasserlöslich, rein Indikator
Bromthymolblau Natriumsalz
Formel: C₂₇H₂₇Br₂NaO₅S
Schmelzpunkt: ? 200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077263
CAS-Nummer: 34722-90-2
EINECS: 252-169-7

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to orange-brown Powder
Infrared spectrum Conforms
UV spectrum Complies
Solubility (0.1 % in water) Clear
Visual transition interval from pH 6.0 (yellow) to pH 7.6 (blue)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR