Phenolrot, rein Indikator

Lieferant: Thermo Fisher Scientific
Achtung

151430250 151430050 151431000
ACRO151430250EA 82.3 EUR
ACRO151430250 ACRO151430050 ACRO151431000
Phenolrot, rein Indikator
Phenolrot
Formel: C₁₉H₁₄O₅S
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003552
CAS-Nummer: 143-74-8
EINECS: 205-609-7
Merck Index: 13,07329

Bestellen

Spezifikation Testergebnisse

Appearance Red Powder
Infrared spectrum Conforms
Solubility (0.1 % in 95% ethanol) Clear soluble
UV Lambda max 555 to 561 nm (in buffer)
Specific extinction E (1%/1cm) ≥1700 (at lambda max., in buffer)
Visual transition interval from pH 6.8 (yellow) to pH 8.2 (red)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR