Myricetin 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
151590250
ACRO151590250EA
92.3
EUR
ACRO151590250
Myricetin 95%
Myricetin
Appearance: Brown-yellow to green-brown Crystalline powder
Formel:
C₁₅H₁₀O₈ Molecular Weight: 318.24 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006827 CAS-Nummer: 529-44-2 |
Spezifikation Testergebnisse
Appearance | Brown-yellow to green-brown Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥94 % |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...