Hydroxyharnstoff 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Hydroxycarbamid, Aminoameisensäurehydroxyamid
151680050
151680250
151681000
ACRO151680050EA
88.5
EUR
ACRO151680050
ACRO151680250
ACRO151681000
Hydroxyharnstoff 98%
Hydroxyharnstoff
Formel:
CH₄N₂O₂ Molecular Weight: 76.05 g/mol Siedepunkt: 296 Schmelzpunkt: 142…146 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00007943 CAS-Nummer: 127-07-1 EINECS: 204-821-7 Merck Index: 13,04874 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 142 to 146 °C |
Titration Iodometric | ≥97.5 % (on dried substance) |
Loss on drying | ≤1 % (1 g, 60 °C) (< 0.7 kPa) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...