4-(Benzyloxy)anilin Hydrochlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

152290250 152291000
ACRO152290250EA 32.9 EUR
ACRO152290250 ACRO152291000
4-(Benzyloxy)anilin Hydrochlorid 98%
4-(Benzyloxy)anilin Hydrochlorid
Formel: C₁₃H₁₄ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00012995
CAS-Nummer: 51388-20-6
EINECS: 257-170-6

Bestellen

Spezifikation Testergebnisse

Appearance Brown Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR